CSM Isolator Sleeves (Two-Piece Glove System)
Actual product may vary
PIERCAN USA
CSM Isolator Sleeves (Two-Piece Glove System)
Chlorosulphonated Polyethylene (CSM) Isolator Sleeve, part of a two-piece system paired with ELS glove. Used in microelectronics, life science, industrial industries; suitable for handling oxidizing products, acids, alkalis and alcohols.
Have Questions?
Catalog NumberPort Size (English)Thickness (English)
FIS17-010-5048 in.15 mil
Catalog number FIS17-010-504
Price (SGD)
1,664.00
Pair
Port Size (English):
8 in.
Thickness (English):
15 mil
Price (SGD)
1,664.00
Pair
  • Two-Piece Glove System with PIERCAN ELS Glove
  • Thickness: 15 mil
  • Length: 24 in.
  • Resistant to Alcohols, Acids, Bases, Strong Oxidants, Uv, Ozone, Free Radicals, and natural aging
  • Resistanat to sterilizations: Autoclave Sterilization, Gamma Radiation (25-50 kG), Peracetic Acid, H2O2 at 35% and 50%
  • Strong resistance to abrasions and impermeable to gas
Specifications
ColorWhite
Latex FreeYes
MaterialCSM
Size8
Thickness (English)15 mil
Thickness (Metric)0.40 mm
For Use With (Application)Industrial and automotive applications
Grip DesignSmooth
Model8Y1524-S
Packaging TypeVacuum-sealed Bag
Port Size (English)8 in.
Port Size (Metric)20 cm
Length (English)24 in.
Length (Metric)60 cm
Unit SizePair