CSM Short Isolator Gloves (Two-Piece Glove System)
Actual product may vary
PIERCAN USA
CSM Short Isolator Gloves (Two-Piece Glove System)
Chlorosuphonated Polyethylene (CSM) Short Isolator Glove, part of a two-piece system that connects to a sleeve, has strong resistance to chemical and sterilizing agents, ideal for microelectronics, life science, and industrial industries.
Have Questions?
Catalog NumberSizeThickness (Metric)
FIS17-010-41390.38 mm
Catalog number FIS17-010-413
Price (SGD)
729.00
Pair
Size:
9
Thickness (Metric):
0.38 mm
Price (SGD)
729.00
Pair
  • Two-Piece Glove System with PIERCAN sleeve
  • Thickness: 15 mil
  • Length: 13 in.
  • Hand Size: 6 to 10
  • Ambidextrous
  • Resistant to Alcohols, Acids, Bases, Strong Oxidants, Uv, Ozone, Free Radicals, and natural aging
  • Resistanat to sterilizations: Autoclave Sterilization, Gamma Radiation (25-50 kG), Peracetic Acid, H2O2 at 35% and 50%
  • Strong resistance to abrasions and impermeable to gas
Specifications
ColorWhite
HandAmbidextrous
Latex FreeYes
MaterialCSM
Size9
Temperature (English) Max.260°F
Temperature (Metric) Max126°C
Thickness (English)15 mil
Thickness (Metric)0.38 mm
Grip DesignSmooth
ModelY15-9A
Packaging TypePacked by the pair and heat sealed in 0.3 mm vacuum bags
Length (English)13 in.
Length (Metric)33 cm
Unit SizePair