Neoprene One-Piece Glove, 8 in. Port
Actual product may vary
PIERCAN USA
Neoprene One-Piece Glove, 8 in. Port
Polychloroprene (Neoprene) One-Piece glove has flexibility, dexterity and excellent resistance to sterilizing agents; compatible in the microelectronics, life science, industrial, and nuclear industries.
Have Questions?
Catalog NumberSizeThickness (Metric)Hand
FIS17-010-30590.60 mmAmbidextrous
Catalog number FIS17-010-305
Price (SGD)
13,433.00
Pack
Size:
9
Thickness (Metric):
0.60 mm
Hand:
Ambidextrous
Price (SGD)
13,433.00
Pack
  • One-Piece Glove System
  • Port Size: 8
  • Thickness: 15, 24 or 30 mil
  • Length: 32 in.
  • Hand Size: 8 to 10
  • Ambidextrous
  • Resistant to alcohols, acids, bases, and ketone solvents
  • Resistance to sterilization agents: Paracetic Acid, autoclave sterilization, and H2O2 at 35% and 50%
  • Resistant to abrasions
Specifications
Certifications/Compliance (Europe)Cat 3, EN 421
ColorBlack
HandAmbidextrous
Latex FreeYes
MaterialNeoprene
Size9
SterilityNon-sterile
Temperature (English) Max.212°F
Temperature (Metric) Max100°C
Thickness (English)24 mil
Thickness (Metric)0.60 mm
For Use With (Application)Nuclear industries, life sciences, pharmaceutical
Grip DesignSmooth
Model8N2432-9A
Port Size (English)8 in.
Port Size (Metric)20 cm
Length (English)32 in.
Length (Metric)81.2 cm
Unit SizePack