Check Order Status
Buffer, Reference Standard, pH 12.45 ± 0.01 at 25°C, Ricca Chemical
Structure Search
Buffer, Reference Standard, pH 12.45 ± 0.01 at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS1618-16
500 mL
Clear PET-G Bottle
Catalog number
FIS1618-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Clear PET-G Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 1310-73-2, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Shelf Life
18 months
Name Note
pH 12.45± 0.01 at 25°C
CAS
7447-40-7
CAS Min %
97.0
View more