Check Order Status
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 10.00 ± 0.01 at 25°C (Color Coded Blue), Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS1601-16
500 mL
Natural Poly Bottle
Catalog number
FIS1601-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
497-19-8, 144-55-8, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Name Note
pH 10.00 ± 0.01 at 25 deg C (Color Coded Blue)
CAS
497-19-8
Melting Point
0°C
Boiling Point
100°C
View more