Rotary Vane Vacuum Pumps
Actual product may vary
Labconco
Rotary Vane Vacuum Pumps
Designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets
Have Questions?
Change viewbuttonViewtableView
Catalog NumberPlug TypeFlow RateAmperage
FIS16-315-51NEMA 6-15P117 L/min.2.4 A
LBC7739402Reverse IEC117 L/min.2.4 A
Catalog number FIS16-315-51
Price (SGD)
-
Plug Type:
NEMA 6-15P
Flow Rate:
117 L/min.
Amperage:
2.4 A
Designed for use with Labconco Freeze-dry Systems—compact size easily fits into freeze-dry system cabinets
Provide a reliable vacuum source for other products including glove boxes and centrifugal concentrators.
  • Two-stage, oil-sealed pumps with sliding vane
  • Switchable high-vacuum and high-throughput modes
  • Three-position gas ballast (Closed, Low Flow, and High Flow)
  • Enclosed, fan-cooled, single-phase electric motor drives pump
  • Thermal overload protection with automatic restart
  • Inlet valve prevents air and oil suckback in the event of power or oil loss
  • Cast-aluminum housing
  • Mounted on a base plate with rubber feet for stability
Cat. No. 16-108-218 has a reverse IEC plug; models with 117L/min. pumping capacity include a retractable lifting handle; models with 195L/min. pumping capacity have a lifting bracket. A pump exhaust filter with oil and odor filter elements is also included.
Specifications
Max. Vacuum2 x 10-3 mBar
Amperage2.4 A
IncludesPump, four each one liter bottles of vacuum pump oil and one exhaust filter with oil mist and odor filter elements. ON/OFF switch, power cord with plug for direct connection of pump to Labconco freeze dryers, one 12.7mm (1/2 in.) O.D. and one 19.1mm (3/4 in.) O.D. inlet adapter
Electrical Requirements230 V 50/60 Hz
MaterialCast Aluminum
Plug TypeNEMA 6-15P
Weight (English)65 lb.
Weight (Metric)29.5 kg
Flow Rate117 L/min.
Frequency50/60 Hz
Overall Height (English)10.4 in.
Overall Height (Metric)26.4 cm
Overall Length (English)17 in.
Overall Length (Metric)43.18 cm
Overall Width (English)6.5 in.
Pump TypeRotary Vane Vacuum Pump
Maximum Pressure1 atm
For Use With (Equipment)Freeze dry system