pep4c
pep4c
Actual product may vary
pep4c
pep4c
pep4c
Inactive control peptide for pep2m (Cat. No. 1595)
Have Questions?
Catalog NumberQuantity
FIS159622 mg
Catalog number FIS15962
Price (SGD)
22.00
PK
Add to cart
Quantity:
2 mg
Price (SGD)
22.00
PK
Add to cart
pep4c is an inactive control peptide analog of pep2m, a peptide inhibitor of the interaction between the C-terminus of the GluA2 (AMPA receptor) subunit and N-ethylmaleimide-sensitive fusion protein (NSF).
Active Analog also available.
For Research Use Only
Specifications
Descriptionpep4c
Molecular Weight (g/mol)1146.42
Product Typepep4c
Quantity2 mg
SequenceLys-Arg-Met-Lys-Val-Ala-Lys-Ser-Ala-Gln
Unit SizePK