Check Order Status
Handheld Tape Dispenser
Actual product may vary
Handheld Tape Dispenser
Accommodates various tape widths, on 1 in. cores, for easy dispensing.
Have Questions?
Catalog number
FIS15942
Price (SGD)
46.00
EA
Add to cart
Price (SGD)
46.00
EA
Add to cart
For 0.50, 0.75, or 1 in. wide (1.3, 1.9, or 2.5cm) tapes on 1 in. (2.5cm) core
Plastic construction
Specifications
Product Type
Tape Dispenser
Material
Plastic
For Use With (Equipment)
For 0.5, 0.75, or 1 in. W tapes on 1 in. core
Model
Handheld