Self-Adhesive Label Tape
Self-Adhesive Label Tape
Actual product may vary
Self-Adhesive Label Tape
All-purpose—even sticks to plastics during autoclaving.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (English)Width (English)Includes
FIS1594014 yd.1.5 in.Label Tape Only
FIS1593714 yd.0.5 in.Tape and Dispenser
FIS1593814 yd.0.75 in.Tape and Dispenser
FIS1593560 yd.1 in.Label Tape Only
FIS1593914 yd.1 in.Label Tape Only
Catalog number FIS15940
Price (SGD)
29.00
EA
Add to cart
Length (English):
14 yd.
Width (English):
1.5 in.
Includes:
Label Tape Only
Price (SGD)
29.00
EA
Add to cart
All-purpose—even sticks to plastics during autoclaving
  • Sticks to glass, plastic, paper, and wood from -23° to +121°C (-9° to +250°F)
  • Especially suited for use in autoclave, refrigerator, deep freeze, sterilizer and incubator
  • Easily marked with pencil, pen, ballpoint and most felt-tip markers
  • Removes without residue
  • Oil, water and acid resistant
Specifications
Adhesive TypeSelf-adhesive
ColorWhite
FormatTape
Product TypeSelf Adhesive Label Tape
For Use With (Application)Autoclave, Refrigerator/freezer, Sterilizer, Incubator
IncludesLabel Tape Only
Quantity1 Ea.
Temperature Range (English)-9 to 250°F
Temperature Range (Metric)-23°C to +121°C
Dimensions (LxW)14 yd. x 1.5 in.
Length (English)14 yd.
Length (Metric)13 m
Width (English)1.5 in.
Diameter (English)1 in. I.D.
Unit SizeEA