Biohazard Labels
Biohazard Labels
Actual product may vary
Biohazard Labels
Biohazard Labels
Biohazard Labels
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantityLength (English)Length (Metric)Width (English)
FIS15928250/Pk.5 in.12.7 cm5 in.
FIS15-928250/Pk.5 in.12.7 cm5 in.
FIS15-927500/Pk.3 in.7.62 cm2 in.
FIS15927500/Pk.3 in.7.62 cm2 in.
Catalog number FIS15928
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250/Pk.
Length (English):
5 in.
Length (Metric):
12.7 cm
Width (English):
5 in.
Bright red fluorescent labels display the universal biohazard symbol for easy identification.
  • Hazard communication labels ensure safety and efficiency
  • Identify equipment, containers, rooms, materials, experimental animals or combinations of these items that may contain potentially infectious agents
  • Easy peel-off labels with pressure-sensitive adhesive
Use in accordance with applicable federal and state regulations.
Specifications
Adhesive TypePressure-sensitive
FormatLabel
MaterialPaper
Product TypeBiohazard Warning Tape
For Use With (Application)Biohazard
Quantity250/Pk.
Length (English)5 in.
Length (Metric)12.7 cm
Width (English)5 in.
Unit SizePK