▸
Cell Analysis Products
▸
Microspheres
▸
Dynabeads™ MX Mixer Turret
Dynabeads™ MX Mixer Turret
Dynabeads™ MX Mixer Turret
Have Questions?
Catalog Number
Quantity
FIS15906
1 Unit
Catalog number
FIS15906
Price (SGD)
13.00
Each
Add to cart
Quantity:
1 Unit
Price (SGD)
13.00
Each
Add to cart
Attachment for Dynabeads™ MX Mixer Base (Cat. No. 159-02). Designed to connect the Mixer Base with the 12-tube Mixing Head (Cat. No. 159-03) or 8-tube Mixing Head (Cat. No. 159-04).
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product Type
MX Mixer Turret
For Use With (Equipment)
Dynabeads™ Sample Mixers
Quantity
1 Unit
Shipping Condition
Room Temperature
Unit Size
Each
Contents & Storage
Store at room temperature.