Colored Labeling Tape, Rainbow Pack
Colored Labeling Tape, Rainbow Pack
Actual product may vary
Colored Labeling Tape, Rainbow Pack
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDimensions (LxW)Quantity
FIS15901R14 yd. x 0.5 in.24/Pk.
FIS15-901-10R14 yd. x 1 in.12/Pk.
FIS15-901-5R14 yd. x 0.75 in.16/Cs.
FIS15-901-R14 yd. x 0.5 in.24/Pk.
FIS15-901R14 yd. x 0.5 in.24/Pk.
Catalog number FIS15901R
Price (SGD)
206.00
PK
Add to cart
Dimensions (LxW):
14 yd. x 0.5 in.
Quantity:
24/Pk.
Price (SGD)
206.00
PK
Add to cart
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Available on 14 yd. (13m) rolls. Core size: 1 in.
  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily
Tapes are supplied without dispenser.
Specifications
ColorAssorted
FormatTape
Product TypeColored Label Tape
For Use With (Application)Oil resistant, Waterproof, Acid resistant and excellent for color coding
Includes3 Blue, 3 pink, 3 green, 3 red, 4 yellow, 3 orange, 5 white
Quantity24/Pk.
Temperature Range (English)-10 to +250°F
Temperature Range (Metric)-23°C to +121°C
Dimensions (LxW)14 yd. x 0.5 in.
Length (English)14 yd.
Length (Metric)13 m
Width (English)0.5 in.
Diameter (Metric)1 in. I.D.
Unit SizePK