Check Order Status
Color-Coded Autoclavable Identification Tape
Actual product may vary
Color-Coded Autoclavable Identification Tape
Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.
Have Questions?
Change view
Catalog Number
Color
FIS1590130H
Aluminum
FIS1590130B
Yellow
FIS15-901-30E
Orange
FIS15-901-30D
Red
FIS15-901-30C
Green
FIS15-901-30F
Blue
FIS15-901-30G
Brown
FIS15-901-30K
Black
8 Options
Catalog number
FIS1590130H
Price (SGD)
21.00
EA
Add to cart
Color:
Aluminum
Price (SGD)
21.00
EA
Add to cart
Instrument marking tape for easy identification and color coding items. Even sticks to PTFE.
Vinyl self-stick tapes color-code autoclave items such as surgical instruments for easy identification
Attach securely and safely to any clean surface
Resist water, oil, and acids
Nontoxic
Specifications
Autoclavable
Autoclavable
Color
Aluminum
Format
Tape
Material
Vinyl
Product Type
Identification Tape
For Use With (Application)
Resist Water, Oil and Acids
Quantity
1 Ea.
Length (English)
16 ft.
Length (Metric)
4.8 m
Width (English)
0.37 in.