Recirculating Chiller for Sonicators Accessories: Tubing/Connector Set
Recirculating Chiller for Sonicators Accessories: Tubing/Connector Set
Actual product may vary
Fisherbrand
Recirculating Chiller for Sonicators Accessories: Tubing/Connector Set
Accessories for recirculating chiller
Have Questions?
Catalog number FIS15338422
Price (SGD)
-
Tubing and connector set for use with Qsonica Recirculating Chiller for Sonicators.
Tubing/Connector Set for Cup Horn
  • Quick-connect tubing and fittings (sold separately) are used to attach Chiller (Cat. No. 15-338-421) to ports on the cup horn
  • Allows cold water to recirculate through the ultrasonic horn reservoir
Specifications
Product TypeTubing and Connector Set
For Use With (Application)Ideal for attach chiller to ports on the cup horn
For Use With (Equipment)Q500 and Q700 Sonicator™
Includes2 male fittings, 2 right angle male fittings and 2 female fittings
Unit SizeEA