Magnifying Loupes
Magnifying Loupes
Actual product may vary
Magnifying Loupes
Quickly and easily locked into personal setting using a unique locking knob
Have Questions?
Catalog NumberMagnificationSizeColor
FIS1501-53xSmallBlack
Catalog number FIS1501-5
Price (SGD)
-
Magnification:
3x
Size:
Small
Color:
Black
  • Dual hinge design
  • Locking interpupillary knob
  • Double locking convergence angle
Specifications
ColorBlack
For Use With (Application)For endodontic procedures
Length (English) Focal10 to 13 in.
Length (Metric) Focal25.4 to 33 cm
SizeSmall
Diameter (Englis) Lens2.8 in.
Diameter (Metric) Lens7.11 cm
Magnification3x
Product TypeLoupe, Magnifying
Unit SizeEA