Check Order Status
Radioactive Tape
Actual product may vary
Radioactive Tape
Apply to bottles, boxes, and vials—anything that may be a radioactive material. Easy to write on.
Have Questions?
Change view
Catalog Number
Display Text
FIS15-967
Radiation symbol and caution
FIS15-943
Radiation symbol, caution and printed headings
2 Options
Catalog number
FIS15-967
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Display Text:
Radiation symbol and caution
Apply to bottles, boxes, and vials—anything that may be a radioactive material. Easy to write on.
Yellow tape with bright red imprint with radiation symbol and “CAUTION RADIOACTIVE MATERIAL” message
Also available with printed headings for Isotope, Amount, and Date followed by space for notations to identify specific material
Specifications
Color
Yellow
Format
Tape
Product Type
Radioactive Tape
Display Text
Radiation symbol and caution
For Use With (Application)
Bottles, boxes and vials anything that may be a radioactive material
Quantity
1 Ea.
Dimensions (LxW)
14 yd. x 0.75 in.
Length (English)
14 yd.
Length (Metric)
13 m
Width (English)
0.75 in.