Check Order Status
Labeling Tape
Actual product may vary
Labeling Tape
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Have Questions?
Change view
Catalog Number
Color
Width (English)
FIS15-959
Salmon
0.75 in.
FIS15-951
Yellow
2 in.
FIS15-950
White
2 in.
FIS15-953
Blue
2 in.
FIS15-954
Red
2 in.
FIS15-958
White
1.5 in.
6 Options
Catalog number
FIS15-959
Price (SGD)
9.00
EA
Add to cart
Color:
Salmon
Width (English):
0.75 in.
Price (SGD)
9.00
EA
Add to cart
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Strong, durable and oil-, water- and acid-resistant
Withstands temperature from -23° to +121°C (-10° to +250°F)
Core diameter: 1 in.
Can be written on with pen, pencil, or permanent marker
Will stick to most surfaces including glass, plastic, and metal
Specifications
Color
Salmon
Format
Tape
Product Type
Label Tape
For Use With (Application)
Waterproof, Oil and Acid resistant
Quantity
1 Ea.
Temperature Range (English)
-10 to 250°F
Temperature Range (Metric)
-23°C to +121°C
Dimensions (LxW)
14 yd. x 0.75 in.
Length (English)
14 yd.
Length (Metric)
13 m
Width (English)
0.75 in.
Diameter (English)
1 in.
Diameter (Metric)
2.5 cm