pep2m
pep2m
Actual product may vary
pep2m
pep2m
pep2m
Peptide inhibitor of GluA2 subunit binding to NSF; reduces AMPA currents
Have Questions?
Catalog NumberQuantity
FIS15-9522 mg
Catalog number FIS15-952
Price (SGD)
15.00
EA
Add to cart
Quantity:
2 mg
Price (SGD)
15.00
EA
Add to cart
Peptide inhibitor of the interaction between the C-terminus of the GluA2 (AMPA receptor) subunit and N-ethylmaleimide-sensitive fusion protein (NSF), a protein that regulates AMPA receptor function. Reduces postsynaptic currents in CA1 neurons, AMPA-mediated currents in cultured hippocampal neurons and AMPA receptor surface expression.
Control Peptide also available.
For Research Use Only
Specifications
Descriptionpep2m
Molecular Weight (g/mol)1173.44
Product Typepep2m
Quantity2 mg
SequenceLys-Arg-Met-Lys-Val-Ala-Lys-Asn-Ala-Gln
Unit SizeEA