Multiple-Roll Tape Dispenser
Multiple-Roll Tape Dispenser
Actual product may vary
Multiple-Roll Tape Dispenser
Multiple-Roll Tape Dispenser
Fisherbrand
Multiple-Roll Tape Dispenser
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
Have Questions?
Catalog NumberHoldsFor Use With (Equipment)
FIS15-912Tapes with 1 in. CoreFor dispensing 9.5 W, rolls with 14 yd. tape
Catalog number FIS15-912
Price (SGD)
134.00
EA
Add to cart
Holds:
Tapes with 1 in. Core
For Use With (Equipment):
For dispensing 9.5 W, rolls with 14 yd. tape
Price (SGD)
134.00
EA
Add to cart
A multiple tape dispenser provides a well-organized means of storing a rainbow assortment of color coding tape.
  • For dispensing multiple rolls with a 3 in. dia. (76.2mm) core (14 yd. [13m] of tape)
  • Accommodate 0.50, 0.75, or 3 in. wide (13, 19, or 25mm) tapes for a total dispensing width of 9.50 in. (24cm)
  • Dispensers stay put with a rubbery felt-type material to keep them from sliding
Specifications
Product TypeMultiple-Roll Tape Dispenser
HoldsTapes with 1 in. Core
MaterialMetal
For Use With (Equipment)For dispensing 9.5 W, rolls with 14 yd. tape
Unit SizeEA