Labeling Tape
Labeling Tape
Actual product may vary
Labeling Tape
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorLength (English)Length (Metric)
FIS15-901HAqua14 yd.13 m
FIS15-901-LLavender14 yd.13 m
FIS15-901-MLime Green14 yd.13 m
FIS15-901-NRose14 yd.13 m
FIS15-901-AWhite14 yd.13 m
FIS15-901-BYellow14 yd.13 m
FIS15-901BYellow14 yd.13 m
FIS15-901-CGreen14 yd.13 m
FIS15-901-DPink14 yd.13 m
FIS15-901-FOrange14 yd.13 m
FIS15-901FOrange14 yd.13 m
FIS15-901-GBlue14 yd.13 m
FIS15-901GBlue14 yd.13 m
FIS15-901-HAqua14 yd.13 m
FIS15-901-15LLavender60 yd.55 m
FIS15-901-15NRose60 yd.55 m
FIS15-901-15AWhite60 yd.55 m
FIS15-901-15BYellow60 yd.55 m
FIS15-901-15CGreen60 yd.55 m
FIS15-901-15ERed60 yd.55 m
FIS15-901-15FOrange60 yd.55 m
FIS15-901-15GBlue60 yd.55 m
FIS15-901-15HAqua60 yd.55 m
FIS15-901-15MLime Green60 yd.55 m
FIS15-901-15QViolet60 yd.55 m
Catalog number FIS15-901H
Price (SGD)
60.00
PK
Add to cart
Color:
Aqua
Length (English):
14 yd.
Length (Metric):
13 m
Price (SGD)
60.00
PK
Add to cart
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily
  • Tapes are supplied without dispenser
  • Core Size: 1 in. (14-yd. tape); 3 in. (60-yd. tape)
Specifications
ColorAqua
FormatTape
MaterialPaper
Product TypeColored Label Tape
For Use With (Application)Waterproof, Oil and Acid resistant
Quantity6/Pk.
Temperature Range (English)-10 to 250°F
Temperature Range (Metric)-23°C to +121°C
Dimensions (LxW)14 yds x 0.5 in.
Length (English)14 yd.
Length (Metric)13 m
Width (English)0.5 in.
Width (Metric)13 mm
Unit SizePK