Colored Labeling Tape 0.5 in x 14 yds, 13mm x 13m; Red
Colored Labeling Tape 0.5 in x 14 yds, 13mm x 13m; Red
Actual product may vary
Colored Labeling Tape 0.5 in x 14 yds, 13mm x 13m; Red
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
Have Questions?
Catalog NumberColor
FIS15-901ERed
Catalog number FIS15-901E
Price (SGD)
60.00
PK
Add to cart
Color:
Red
Price (SGD)
60.00
PK
Add to cart
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily
  • Tapes are supplied without dispenser
Specifications
ColorRed
FormatTape
MaterialPaper
Product TypeColored Label Tape
For Use With (Application)Waterproof, Oil and Acid resistant
Quantity6 Pack
Temperature Range (English)-10 to 250°F
Temperature Range (Metric)-23°C to +121°C
Dimensions (LxW)14 yds x 0.5 in.
Length (English)14 yd.
Length (Metric)13 m
Width (English)0.5 in.
Width (Metric)13 mm
Unit SizePK