Colored Labeling Tape, 1 in. wide
Actual product may vary
Colored Labeling Tape, 1 in. wide
Useful for many labeling applications—will adhere to most surfaces and can be written on.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberColorLength (English)
FIS15-901-10QViolet14 yd.
FIS15-901-10JCopper14 yd.
FIS15-901-10BYellow14 yd.
FIS15-901-10AWhite14 yd.
FIS15-901-10CGreen14 yd.
FIS15-901-10DPink14 yd.
FIS15-901-10ERed14 yd.
FIS15-901-10FOrange14 yd.
FIS15-901-10KGray14 yd.
FIS15-901-10HAqua14 yd.
FIS15-901-10GBlue14 yd.
FIS15-901-10MLime Green14 yd.
FIS15-901-25DPink60 yd.
FIS15-901-25HAqua60 yd.
FIS15-901-25LLavender60 yd.
FIS15-901-25MLime Green60 yd.
FIS15-901-25NRose60 yd.
FIS15-901-25AWhite60 yd.
FIS15-901-25GBlue60 yd.
FIS15-901-25BYellow60 yd.
FIS15-901-25CGreen60 yd.
FIS15-901-25ERed60 yd.
FIS15-901-25FOrange60 yd.
Catalog number FIS15-901-10Q
Price (SGD)
19.00
Pack
Color:
Violet
Length (English):
14 yd.
Price (SGD)
19.00
Pack
Colored Label Tape is useful for many labeling applications. It can be written on and will adhere to most surfaces.
  • Strong, durable, and oil-, water-, and acid-resistant
  • Excellent for color coding
  • Withstand temperatures from -23° to +121°C (-10° to +250°F)
  • Can be written on with pen, pencil, or permanent marker
  • Stick to most surfaces including glass, plastic, and metal
  • Remove cleanly and easily
  • Width is 1 inch
Note: 14 yd. (13m) rolls have a 1 in. core; 60 yd. (55m) rolls have a 3 in. core.
Tapes are supplied without dispenser
Specifications
ColorViolet
FormatTape
MaterialPaper
Product TypeColored Label Tape
For Use With (Application)Waterproof, Oil and Acid resistant
Quantity3/Pk.
Temperature Range (English)-10° to 250°F
Temperature Range (Metric)-23° to +121°C
Dimensions (L x W)14 yd. x 1 in.
Length (English)14 yd.
Length (Metric)13 m
Width (English)1 in.
Width (Metric)25 mm
Unit SizePack