Platform for Microprocessor Controlled Orbital Shaker
Actual product may vary
Boekel Scientific
Platform for Microprocessor Controlled Orbital Shaker
This is an interchangeable platform for Boekel Orbital Shakers (Cat. No. 15-600-317).
Have Questions?
Catalog NumberHolds
FIS15-600-320Flasks and Beakers of All Sizes
Catalog number FIS15-600-320
Price (SGD)
691.00
Each
Holds:
Flasks and Beakers of All Sizes
Price (SGD)
691.00
Each
  • Universal Platform with Adjustable Bars
  • Adjust Bars to Fit Flasks and Beakers of All Sizes
  • interchangeable Locking Design
Specifications
Load Bearing Capacity (Metric)2.5 kg
HoldsFlasks and Beakers of All Sizes
Product TypeUniversal Platform with Adjustable Bars
Length (English)10.6 in.
Length (Metric)270 mm
Width (English)7.9 in.
Width (Metric)200 mm
For Use With (Equipment)Boekel Microprocessor Controlled Orbital Shaker
SizeUniversal
Unit SizeEach