Hydrion™ Sanitizer: Quaternary Test Paper
Actual product may vary
Micro Essential Laboratory
Hydrion™ Sanitizer: Quaternary Test Paper
Provides a simple, reliable, and economical means to measure quarternary concentration of ammonium, chlorine and iodine sanitizer solutions
Have Questions?
Catalog NumberQuantity
FIS15-420-130150 Tests
Catalog number FIS15-420-130
Price (SGD)
190.00
Case
Quantity:
150 Tests
Price (SGD)
190.00
Case
Provides a simple, reliable, and economical means to measure quarternary concentration of ammonium, chlorine and iodine sanitizer solutions
Federal, State and Local health regulations require users of Quaternary Ammonium Sanitizer Solutions to have appropriate test kits available to verify the strength of sanitizer solutions. Sanitizer solutions are essential in the food service industry to ensure that sanitizers are at the proper concentration specified by the individual manufacturer.
High Range Quat Test (FS Cat. No. 15-420-130)
  • Measures high range concentrations
  • Color matches at 0-200-400-600-800-100ppm
  • 15 ft. (4.6m) roll
  • 150 tests per roll
Specifications
For Use With (Application)To measure the concentration of quaternary sanitizers, specifically alkyldimethylbenzyl ammonium compounds, Rocall II and Hyamine 3500
IncludesOne 15 ft. roll of test paper, matching color chart
Quantity150 Tests
TypeQuaternary Test Paper
Unit SizeCase