PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Actual product may vary
HF Scientific
PPD-2 DPD Powder Pop™ Dispensers for Chlorine Testing
Use with any DPD-based photometer or test kit
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Application)For Use With (Equipment)Volume (Metric)
FIS15-393-64Total Chlorine TestingHF Scientific, Hach5 mL
FIS15-393-71Total Chlorine TestingNon-HF, Hach or Hanna10 mL
FIS15-393-65Chlorine TestingHF Scientific, Hach5 mL
Catalog number FIS15-393-64
Price (SGD)
-
For Use With (Application):
Total Chlorine Testing
For Use With (Equipment):
HF Scientific, Hach
Volume (Metric):
5 mL
Portable dispenser accurately delivers the correct volume of DPD reagent for Free or Total Chlorine—use with any DPD-based photometer or test kit.
  • Supplied with enough DPD reagent for 400 tests
  • To use, simply invert dispenser over sample and press button to dispense reagent; powder dissolves quickly in sample, leaving no messy residue behind
  • Available for 5 and 10mL sample sizes, with formulations for a variety of photometers and test kits
Specifications
Volume (Metric)5 mL
For Use With (Application)Total Chlorine Testing
For Use With (Equipment)HF Scientific, Hach
IncludesPocket photometer
Quantity400 Tests
TypeChlorine Reagent Powder Pop Dispenser
Volume (English)0.16 oz.
Unit SizeEach