Procal Primary Calibration Kit
Actual product may vary
HF Scientific
Procal Primary Calibration Kit
Have Questions?
Catalog NumberTurbidity
FIS15-393-560.02, 10, 1000 NTU
Catalog number FIS15-393-56
Price (SGD)
-
Turbidity:
0.02, 10, 1000 NTU
Specifications
TypePrimary Calibration Kit
For Use With (Equipment)MicroTPI and MicroTPW Portable Turbidimeters
Turbidity0.02, 10, 1000 NTU
Unit SizeEach