Enterprise Services
Vendor Managed Inventory Services
Brands
Shop by Brand
eSolutions
Offers
Webinars
Chemical Structure Search
Support
Fisher Scientific Home
Search
Fisher Scientific
Quick Order
Offers
Sign In
Account options
See All Product Categories
Shop by Brand
Browse All Brands
Services
Enterprise Services
eSolutions
Vendor Managed Inventory Services
Chemical Structure Search
Support
Webinars
Home
▸
Shop All Products
▸
Glassware Washers and Cleaners
▸
Washer and Cleaner Accessories
▸
Ultrasonic Cleaning Bath Accessory, Covers
Actual product may vary
Branson Ultrasonics
Ultrasonic Cleaning Bath Accessory, Covers
For use with Bransonic cleaning baths
Have Questions?
Catalog Number
Product Type
Material
For Use With (Equipment)
FIS15-345-128
Lid
Plastic
Ultrasonic Bath, Model 3800
Catalog number
FIS15-345-128
Price (SGD)
142.00
Each
Add to cart
Product Type:
Lid
Material:
Plastic
For Use With (Equipment):
Ultrasonic Bath, Model 3800
Price (SGD)
142.00
Each
Add to cart
For use with Bransonic cleaning baths
Stainless steel lid
Specifications
Product Type
Lid
For Use With (Equipment)
Ultrasonic Bath, Model 3800
For Use With (Application)
Helps position beakers securely in bath
Material
Plastic