Cleaning Bath Accessory, Mesh Baskets
Actual product may vary
Branson Ultrasonics
Cleaning Bath Accessory, Mesh Baskets
For use with Bransonic ultrasonic cleaners
Have Questions?
Catalog NumberCapacity (English)Length (English)Width (English)For Use With (Equipment)
FIS15-337-21B95.99 oz.8 in.4 in.Ultrasonic Bath, Model 2800
Catalog number FIS15-337-21B
Price (SGD)
353.00
Each
Capacity (English):
95.99 oz.
Length (English):
8 in.
Width (English):
4 in.
For Use With (Equipment):
Ultrasonic Bath, Model 2800
Price (SGD)
353.00
Each
For use with Bransonic ultrasonic cleaners
  • Capacity: 0.5 to 1.5 gal.
Specifications
Product TypeMesh Basket
Depth (English)3 in.
Capacity (Metric)2.83 L
Capacity (English)95.99 oz.
Depth (Metric)7.62 cm
For Use With (Equipment)Ultrasonic Bath, Model 2800
For Use With (Application)Ideal for cleaning small parts
Length (English)8 in.
Length (Metric)20.32 cm
MaterialStainless Steel
Dimensions (L x W)8 x 4 in. (20.32 x 10.16 cm)
Width (English)4 in.
Width (Metric)10.16 cm
Unit SizeEach