Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
Actual product may vary
Branson Ultrasonics
Bransonic™ Ultrasonic Cleaning Bath Accessory, Perforated Trays
For use with Branson ultrasonic baths
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)For Use With (Equipment)
FIS15-337-20A25 LUltrasonic Bath, Model 8800
FIS15-337-19C6.8 LUltrasonic Bath, Model 3800
FIS15-337-19B3.4 LUltrasonic Bath, Model 2800
FIS22-06628211.4 LUltrasonic Bath, Model 5800
Catalog number FIS15-337-20A
Price (SGD)
917.00
Each
Capacity (Metric):
25 L
For Use With (Equipment):
Ultrasonic Bath, Model 8800
Price (SGD)
917.00
Each
For use with Branson ultrasonic baths
  • Accessories for ultrasonic bench-top system
Specifications
Product TypePerforated Tray
Capacity (Metric)25 L
Capacity (English)5.5 gal.
For Use With (Equipment)Ultrasonic Bath, Model 8800
MaterialStainless Steel
Dimensions (L x W x H)44.45 x 24.1 x 15.2 cm
Unit SizeEach