CPX Series Ultrasonic Cleaning Bath
Actual product may vary
Branson Ultrasonics
CPX Series Ultrasonic Cleaning Bath
Outfit your lab with a classic tabletop cleaner, offering high-tech design, solid construction and featuring reliable digital controls with power tracking capabilities.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelCapacity (Metric)Dimensions (D x W x H) InteriorVoltage
FIS15-336-133CPX58009.5 L29 x 24 x 15 cm (11.5 x 9.5 x 6 in.)120 V
FIS15-336-135CPX1800-E1.9 L15 x 14 x 10 cm (6 x 5.5 x 4 in.)230/240 V
FIS15-336-131CPX28002.8 L23 x 14 x 10 cm (9.5 x 5.5 x 4 in.)120 V
FIS15-336-132CPX38005.7 L29 x 15 x 15 cm (11.5 x 6 x 6 in.)120 V
FIS15-336-137CPX3800-E5.7 L29 x 15 x 15 cm (11.5 x 6 x 6 in.)230/240 V
FIS15-336-138CPX5800-E9.5 L29 x 24 x 15 cm (11.5 x 9.5 x 6 in.)230/240 V
Catalog number FIS15-336-133
Price (SGD)
5,075.00
Each
Model:
CPX5800
Capacity (Metric):
9.5 L
Dimensions (D x W x H) Interior:
29 x 24 x 15 cm (11.5 x 9.5 x 6 in.)
Voltage:
120 V
Price (SGD)
5,075.00
Each
Outfit your lab with a classic tabletop cleaner, offering high-tech design, solid construction and featuring reliable digital controls with power tracking capabilities.
Bransonic CPX ultrasonic bath cleaners feature digital controls with power tracking capabilities to adjust for light or heavy loads, high/low power control, degassing and sleep mode.
  • Sweep frequency eliminates standing waves and creates consistent cavitation throughout tank
  • 40kHz rugged industrial transducers
  • 99 min. digital timer with continuous ultrasonic operation
  • With built-in pour drain, tank cover
  • Plastic body, chemical-resistant
Ultrasonic Power Tracking
  • Immediately brings tank's frequency back to its optimum point
  • Control circuitry actively tracks operating frequency of bath
  • Delivers unwavering cavitation, steady activity in bath even when load conditions change
Degassing and Conditioning the Solution
  • Can be set up to 99 minutes
  • Extended degas time periods allow beyond cleaning applications such as sample preparation, which includes degassing liquids, mixing and homogenization, dissolving solids, cell lysing, and dispersion of particles
High/Low Power Control
  • Allows user to lower amplitude of acoustic energy in bath: 100% power to tank for normal applications, 70% power for delicate applications
  • Helps to avoid damage to components in bath during ultrasonic process
Sleep Mode
  • An energy-saving feature
  • When cycle is complete and no control keys are touched for 15 minutes, unit goes into sleep mode
  • Touching any key on controller causes unit to come back on
  • Controls are located above and behind cleaning tank in an easily accessible control tower panel, and raised above the tank to avoid damage from cleaning solutions
Specifications
MaterialPlastic
Timer90 min.
Frequency40 kHz
Voltage120 V
Capacity (English)2.5 gal.
Capacity (Metric)9.5 L
Dimensions (D x W x H) Interior29 x 24 x 15 cm (11.5 x 9.5 x 6 in.)
For Use With (Application)Cleaning
IncludesTank cover and drain
ModelCPX5800
Weight (English)14 lb.
Weight (Metric)6.4 kg
Unit SizeEach