12.7 mm Cryogenic Color Dots for Microtube Tops
Actual product may vary
Branson Ultrasonics
12.7 mm Cryogenic Color Dots for Microtube Tops
Writable color dots supplied in sheets, available in a wide range of colors.
Have Questions?
Catalog NumberColor
FIS15-336-124Mint
Catalog number FIS15-336-124
Price (SGD)
9,385.00
Each
Color:
Mint
Price (SGD)
9,385.00
Each
  • 12.7 mm labels available in a wide range of colors for color coding and easy identification
  • Small round labels are ideal for 0.5 to 1.5 mL Eppendorf tube caps and vials
  • Strong adhesive ensures labels won't wrinkle or peel during long-term storage in -80°C freezers, dry ice, and liquid nitrogen
  • Resistant to gamma irradiation (tested up to 50 KGy)
Specifications
Adhesive TypePermanent
ColorMint
FinishMedium Gloss
FormatSheet
MaterialThermoplastic Film
Product TypeLabel
SterilityNon-sterile
Labels per Sheet60/Sheet
No. per Pack7/Pk.
Quantity420/Pk.
ShapeRound
Temperature Range (English)-321°F to +230°F
Temperature Range (Metric)-196°C to +110°C
Dimensions (L x W)12.7 mm
Unit SizeEach