M Series Ultrasonic Cleaning Bath
Actual product may vary
Branson Ultrasonics
M Series Ultrasonic Cleaning Bath
Outfit your lab with a Bransonic MH Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberModelVoltageCapacity (Metric)Dimensions (D x W x H) Interior
FIS15-336-101M2800120 V2.8 L23 x 14 x 10 cm (9.5 x 5.5 x 4 in.)
FIS15-336-105M1800-E230/240 V1.9 L15 x 14 x 10 cm (6 x 5.5 x 4 in.)
FIS15-336-106M2800-E230/240 V2.8 L23 x 14 x 10 cm (9.5 x 5.5 x 4 in.)
FIS15-336-109M8800-E230/240 V20.8 L49.5 x 29 x 15 cm (19.5 x 11.5 x 6 in.)
Catalog number FIS15-336-101
Price (SGD)
2,294.00
Each
Model:
M2800
Voltage:
120 V
Capacity (Metric):
2.8 L
Dimensions (D x W x H) Interior:
23 x 14 x 10 cm (9.5 x 5.5 x 4 in.)
Price (SGD)
2,294.00
Each
Outfit your lab with a Bransonic MH Series Ultrasonic Cleaner. This unit is designed with solid construction and excellent functionality for a variety of applications − including cell separation, sample preparation and degassing of liquids.
  • Sweep frequency eliminates standing waves and creates consistent cavitation throughout the tank, eliminating dead zones for long life
  • Includes 60-minute mechanical timer and continuous operation
  • 40kHz industrial transducers
  • Controls located above and behind in easily accessible control tower panel which is raised above tank to avoid damage from cleaning solutions
  • Plastic body
  • Chemical-resistant
  • 1.5 gal. and larger models
  • Tank drains with valves
Specifications
MaterialPlastic Housing, Stainless Steel Tank
Timer60 min.
Frequency40 kHz
Voltage120 V
Capacity (English)0.75 gal.
Capacity (Metric)2.8 L
Dimensions (D x W x H) Interior23 x 14 x 10 cm (9.5 x 5.5 x 4 in.)
IncludesTank cover, mechanical timer
ModelM2800
Weight (English)9 lb.
Weight (Metric)4.1 kg
Unit SizeEach