Polypropylene Tubing T-Shaped Connectors
Actual product may vary
Polypropylene Tubing T-Shaped Connectors
Withstand flows to 20psi.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberDiameter (English) InnerHeight (English)Overall Length (English)Width (English)
FIS15-315-26A0.156 in.0.58 in.0.15 in.0.8 in.
FIS15-315-26B0.218 in.0.9 in.1.185 in.0.2 in.
FIS15-315-26C0.312 in.1.2 in.1.5 in.0.29 in.
Catalog number FIS15-315-26A
Price (SGD)
249.00
Case
Diameter (English) Inner:
0.156 in.
Height (English):
0.58 in.
Overall Length (English):
0.15 in.
Width (English):
0.8 in.
Price (SGD)
249.00
Case
  • Excellent chemical resistance
  • Polypropylene
  • Autoclavable
  •  Serrated connecting surfaces to prevent tubing slipping
  • Withstands flows to 20 p.s.i.
Specifications
AutoclavableAutoclavable
Diameter (English) Inner0.156 in.
MaterialPolypropylene
Overall Length (English)0.15 in.
Overall Length (Metric)3.81 mm
For Use With4mm
Width (Metric)20.32 mm
Width (English)0.8 in.
ShapeT
Height (Metric)14.7 mm
Height (English)0.58 in.
ColorTransparent
Maximum Pressure20 psi
Product TypeTubing Connector
Unit SizeCase