Check Order Status
Filter Disc, Untreated, Ground Cylindrical, 25 mm
Actual product may vary
Thermo Scientific
Filter Disc, Untreated, Ground Cylindrical, 25 mm
The faces of the VitraPOR™ standard filter discs are press moulded and may show thickness deviations.
Have Questions?
Catalog Number
Porosity
FIS15-251
1
Catalog number
FIS15-251
Price (SGD)
146.00
EA
Add to cart
Porosity:
1
Price (SGD)
146.00
EA
Add to cart
Highly resistant to most chemicals and temperatures up to 500°C
Specifications
Material
Borosilicate Glass (3.3)
Pore Size
1 μm
Sterility
Non-sterile
No. per Pack
10/Pk.
Porosity
1
Thickness (Metric)
3.0 ± 0.7 mm
Diameter (Metric)
25 mm