Polypropylene Sterilizing Trays with Cover
Polypropylene Sterilizing Trays with Cover
Actual product may vary
Bel-Art
Polypropylene Sterilizing Trays with Cover
Instrument trays for sterilizing
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS15-249C11 x 6 x 5 in.
Catalog number FIS15-249C
Price (SGD)
177.00
EA
Add to cart
Dimensions (L x W x H):
11 x 6 x 5 in.
Price (SGD)
177.00
EA
Add to cart
Instrument trays for sterilizing
  • Smooth, rounded corners for easy cleaning and sterilizing
  • Outward tapered walls allow nesting for compact storage
  • Covers are inset into tray for stable stacking of loaded trays
  • Steam autoclavable at 121°C (250°F)
Specifications
Capacity (Metric)5385 cm3
Dimensions (L x W)11 x 6 in. (29 x 15 cm)
Dimensions (L x W x H)11 x 6 x 5 in.
ColorNatural
Height (Metric)12.7 cm
Length (Metric)27.9 cm
MaterialPolypropylene
TypeInstrument Tray
Width (Metric)15.2 cm
IncludesCover
Length (English)11 in.
Height (English)5 in.
ShapeRectangular
Width (English)6 in.