Chemical-Resistant Multipurpose Trays and Pans
Actual product may vary
MFG Tray
Chemical-Resistant Multipurpose Trays and Pans
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Have Questions?
Catalog NumberDimensions (L x W x H)Type
FIS15-239-1624 x 12 x 2.75 in.Multi-purpose Tray and Ventilated Stacking Pan
Catalog number FIS15-239-16
Price (SGD)
98.00
Each
Dimensions (L x W x H):
24 x 12 x 2.75 in.
Type:
Multi-purpose Tray and Ventilated Stacking Pan
Price (SGD)
98.00
Each
Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
  • Reinforced white thermoset polyester composite construction Resistant to most detergents, mild acid and alkaline solutions, corrosive salts and many reagents
  • Withstand temperatures from -51° to +121°C (-60° to +250°F) continuous exposure, or 149°C (300°F) intermittent
  • High load-bearing capacity
  • Excellent dimensional stability Easily steam cleaned
Specifications
Dimensions (L x W x H)24 x 12 x 2.75 in.
ColorWhite
Height (Metric)7 cm
Length (Metric)61 cm
MaterialFiberglass
TypeMulti-purpose Tray and Ventilated Stacking Pan
Width (Metric)30.5 cm
For Use With (Application)Ideal for soaking, cleaning, autoclaving, storing or carrying lab products and containing spills
Length (English)24 in.
Height (English)2.75 in.
Temperature (English)-60°F to +250°F
Temperature (Metric)-51°C to +121°C
Width (English)12 in.
Unit SizeEach