Specifications
Product TypeCleaning Cloth
Length (English)13.4 in.
Length (Metric)34 cm
Width (English)13.38 in.
Width (Metric)24.89 cm
MaterialNon-woven Polyproylene Cellulose
ColorWhite
Dimensions (L x W)13.4 x 9.8 in.
For Use With (Application)Extra scrubbing and cleaning power
Unit SizeCase