Chex-All™ Heat Sealable Sterilization Pouches
Actual product may vary
Chex-All™ Heat Sealable Sterilization Pouches
Chex-All™ Heat Sealable Sterilization Pouches
Propper
Chex-All™ Heat Sealable Sterilization Pouches
Heat-Sealable Chex-All™ Sterilization Pouches are premium quality sterile packaging products designed to reduce sterilization processing costs. For use in steam and ethylene oxide. Pre-printed indicator inside and outside the pouch. Triple-sealed paper/plastic film ensures content security.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberLength (Metric)Width (Metric)
FIS15-234-330.5 cm17.8 cm
FIS15-234-561 cm45.7 cm
Catalog number FIS15-234-3
Price (SGD)
1,101.00
Pack
Length (Metric):
30.5 cm
Width (Metric):
17.8 cm
Price (SGD)
1,101.00
Pack
Heat-sealable
Chex-All™ Heat Seal Sterilization Pouches provide a sterile barrier, and feature easy-to-read pre-printed indicators (both inside and outside the pouch). Heat-Sealing pouches provide the convenience of custom sizing to meet the individual needs of your instruments.
  • Chex-All™ pouches are for use in steam and ethylene oxide, eliminating the need for two sets of pouches.
  • Chex-All™ pouches are validated for double pouch packaging.
  • Heat Seal Sterilization Pouches allow you to choose the exact length your need.
  • Easy-to-read chemical indicator changes from yellow to black in steam.
  • Easy-to-read chemical indicator changes from red to green in EO.
  • Made of 2.5 mm plastic and 73-74 gram.
  • Triple sealed paper/plastic film provides excellent sterility protection.
Specifications
AutoclavableAutoclavable
Closure TypeHeat Seal
ColorWhite
DisposableSingle-use
For Use With (Application)Indicator imprints on the inside and outside for both steam and ethylene oxide gas
MaterialPaper, Plastic
SterilityNon-sterile
No. per Case1000/Cs.
Thickness (English)2.50 mil
Dimensions (L x W)12 x 7 in. (30.5 x 17.8 cm)
Length (English)12 in.
Width (English)7 in.
Product TypeSterilization Pouch
Length (Metric)30.5 cm
Width (Metric)17.8 cm
Thickness (Metric)0.064 mm
Unit SizePack