AutoclavableAutoclavable
Closure TypeHeat Seal
ColorWhite
DisposableSingle-use
For Use With (Application)Steam and gas sterilization
MaterialPaper, Plastic
SterilityNon-sterile
No. per Case6/Cs.
Length (English)100 ft.
Width (English)9 in.
Product TypeSterilization Reel
Length (Metric)30.5 m
Width (Metric)228.6 mm
Unit SizeEach