General Purpose Crucible Tongs
General Purpose Crucible Tongs
Actual product may vary
Fisherbrand
General Purpose Crucible Tongs
Fisherbrand General Purpose Crucible Tongs are made from ASTM™ standard stainless-steel tongs with nonbinding joints
Have Questions?
Catalog NumberLength (Metric)
FIS15-19323 cm
Catalog number FIS15-193
Price (SGD)
67.00
EA
Add to cart
Length (Metric):
23 cm
Price (SGD)
67.00
EA
Add to cart
Fisherbrand General Purpose Crucible Tongs are made from ASTM™ standard stainless-steel tongs with nonbinding joints
Fisherbrand™ Economical Crucible Tongs are designed to exceed performance expectations -This is the Fisherbrand Advantage.
  • Accurately fabricated parts are flattened at the joint
  • Fastened with a spun (not hammered) rivet
  • Large finger openings facilitate firm grip and good control of tongs
  • Corrosion resistant
Specifications
IncludesFastened with a spun rivet, large finger openings
Length (English)9 in.
Surface CoatingCorrosion Resistant
Length (Metric)23 cm
MaterialStainless Steel
TypeCrucible Tongs
Unit SizeEA