Search Thermo Fisher Scientific
| Catalog Number | Capacity (Metric) | Closure Type | Closure Color | Label |
|---|---|---|---|---|
| FIS15-182-77 | 2 L | Snap Cap | White | In case |
| FIS15-182-78 | 5 L | Snap Cap | White | In case |
| FIS15-182-84 | 250 mL | Screw Cap with O-Ring | Yellow | In case |
| FIS22-045-174 | 120 mL | Screw Cap with O-Ring | Yellow | In case |