GIANT-DIGITS™ Radio Atomic Traceable Clock with Humidity
Actual product may vary
GIANT-DIGITS™ Radio Atomic Traceable Clock with Humidity
GIANT-DIGITS™ Radio Atomic Traceable Clock with Humidity
GIANT-DIGITS™ Radio Atomic Traceable Clock with Humidity
Allows users to view time, temperature and humidity from anywhere in the lab
Have Questions?
Catalog NumberTemperature Range (Metric)
FIS15-079-6800°C to 45°C
Catalog number FIS15-079-680
Price (SGD)
419.00
Each
Temperature Range (Metric):
0°C to 45°C
Price (SGD)
419.00
Each
  • Giant-Digits™ 4-1/2 inches viewable from 200 feet
  • Radio signal from NIST's Colorado atomic clock automatically sets correct date/time
  • Calendar displays month/date, day, leap year, alarm, plus temperature and humidity
  • Temperature range: 0 to 45°C (32 to 113°F); resolution: 0.1°; accuracy: ±1°C
  • Humidity range: 20 to 90% RH; resolution: 0.1°; accuracy: ±5% mid-range (±7% otherwise)
  • Clock automatically adjusts for daylight savings or leap years
  • Desk stand and wall mount supplied
  • Simultaneously displays both F/C temperatures and relative humidity
  • 12/24 hour format
  • Resolution is 1 second
  • Accuracy is 1/10,000 of a second
  • Maintains better than 1-second accuracy for 25,000 years
Specifications
TypeWall Clock
Humidity Range20 to 90%
Resolution0.1°
Accuracy±5% mid-range (±7% otherwise)
Temperature Range (Metric)0°C to 45°C
Temperature Accuracy±1°C
Alarm TypesTime of Day Alarm
Battery Type4 x AA
Display TypeLCD
Dimensions (L x W x H)17 x 1.3 x 11.3 in.
Length (Metric)43.2 cm
Width (Metric)3.3 cm
Height (Metric)28.7 cm
Weight (Metric)0.45 kg
IncludesBatteries, Wall mount, Bench stand, Traceable Certificate
Certifications/ComplianceISO 17025, A2LA, NIST
Unit SizeEach