Digital Thermometers with Stainless-Steel Stem and 0.375 in. LCD Screen
Digital Thermometers with Stainless-Steel Stem and 0.375 in. LCD Screen
Actual product may vary
Digital Thermometers with Stainless-Steel Stem and 0.375 in. LCD Screen
Easy-to-read handheld digital thermometers
Have Questions?
Change viewbuttonViewtableView
Catalog NumberAccuracyCertifications/Compliance
FIS15-078L±1°CISO 17025, A2LA, NIST
CTL14-648-47±0.3°CDNV, ISO 17025, A2LA
CTL15-078-L±1°CISO 17025, A2LA, NIST
CTL15-078L±1°CISO 17025, A2LA, NIST
Catalog number FIS15-078L
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Accuracy:
±1°C
Certifications/Compliance:
ISO 17025, A2LA, NIST
Easy-to-read Fisher Scientific™ Digital Thermometers with Stainless-Steel Stem are chemically resistant to acids, bases, solvents, and may be used to measure temperature of gases, food, and other semisolids.
  • Short-stem piercing models are ideal for carrying in a pocket
  • Longer probes are excellent for measuring sample temperatures while stirring in deep vessels or water baths
  • Thermometers with longer sampling rates allow thermometer to be removed from sample before reading - ideal when display is difficult to read in measuring position
  • For use in cuvets, soil, incubators, petroleum products and foods
Specifications
Certifications/ComplianceISO 17025, A2LA, NIST
Diameter (English) Probe0.15 in.
For Use With (Application)Measuring temperatures of semi solids gunky and viscous solutions
IncludesSilver Oxide battery, Stainless Steel probe
Length (English) Probe5.25 in.
Length (Metric)165 mm
Length (Metric) Probe13.3 cm
MaterialPlastic
Resolution0.1°C
Weight (English)1 oz.
Weight (Metric)28 g
Product TypeDigital Thermometer
Length (English)6.5 in.
Max. Temperature (English)302°F
Max. Temperature (Metric)150°C
Min. Temperature (English)-58°F
Min. Temperature (Metric)-50°C
Temperature Range (English)-58°F to +302°F
Temperature Range (Metric)-50°C to +150°C
CalibratedYes
Accuracy±1°C
DisplayDigital LCD
Width (Metric)8 cm
Unit SizeCS