LED Magnifying Lamp
LED Magnifying Lamp
Actual product may vary
LED Magnifying Lamp
Useful in inspection, electronics, quality control, sample examination, and biological analysis.
Have Questions?
Catalog NumberMagnification
FIS15-078-2203x
Catalog number FIS15-078-220
Price (SGD)
328.00
EA
Add to cart
Magnification:
3x
Price (SGD)
328.00
EA
Add to cart
Useful in inspection, electronics, quality control, sample examination, and biological analysis.
  • Brilliant LED light closely resembles daylight for optimal color detection and clarity
  • Large viewing area (4 x 2.5 in.) is easy to read and arm extension is 31 inches
  • Clamps vertically or horizontally to top or side of any edge
Specifications
Description4 x 2.5 in. magnification lens
IncludesA/C plug, bench clamp
Diameter (Englis) Lens4 x 2.5 in.
Magnification3x
Unit SizeEA