Check Order Status
Buffer, Reference Standard, pH 1.68 ± 0.01 at 25°C, Ricca Chemical
Element Search
Structure Search
Buffer, Reference Standard, pH 1.68 ± 0.01 at 25°C, Ricca Chemical
Have Questions?
Catalog Number
Quantity
Packaging
FIS1492-16
500 mL
Natural Poly Bottle
Catalog number
FIS1492-16
Price (SGD)
-
Quantity:
500 mL
Packaging:
Natural Poly Bottle
Request bulk or custom format
Chemical Identifiers
CAS
7447-40-7, 7732-18-5
Specifications
Chemical Name or Material
Buffer, Reference Standard
Shelf Life
24 months
Name Note
pH 1.68± 0.01 at 25°C
CAS
7447-40-7
Melting Point
0°C
View more