Mithramycin A
Mithramycin A
Mithramycin A
Mithramycin A
Inhibitor of DNA and RNA polymerase
Have Questions?
Catalog NumberQuantity
FIS1489-11 mg
Catalog number FIS1489-1
Price (SGD)
-
Quantity:
1 mg
Request bulk or custom format
Specifications
Chemical Name or MaterialMithramycin A
CAS18378-89-7
Recommended StorageDesiccate at -20°C
Quantity1 mg
SolubilitySoluble to 50mM in DMSO
View more
Mithramycin A is an anticancer antibiotic that selectively binds to G-C-rich DNA in the presence of Mg2+ or Zn2+, inhibiting RNA and DNA polymerase action. Inhibits c-myc expression and induces myeloid differentiation of HL-60 promyelocytic leukemia cells.