Check Order Status
Buffer Solution, pH 3.5 R, Ricca Chemical
Element Search
Structure Search
Buffer Solution, pH 3.5 R, Ricca Chemical
Have Questions?
Catalog Number
Quantity
FIS1474-16
500 mL
Catalog number
FIS1474-16
View Price:
Sign In
Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS
7647-01-0, 631-61-8
Specifications
Chemical Name or Material
Buffer Solution
CAS Min %
65.0
CAS Max %
68.0
CAS
7647-01-0
CAS Min %
16.53
View more