PYREX™ Filling Bells
PYREX™ Filling Bells
Actual product may vary
PYREX™ Filling Bells
For aseptically filling culture tubes and bottles
Have Questions?
Catalog NumberDiameter (Metric)Height (Metric)
FIS14-983B11 mm85 mm
Catalog number FIS14-983B
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Diameter (Metric):
11 mm
Height (Metric):
85 mm
For aseptic filling of vessels. Rubber tubing is attached to the top of the tubulation while the receiving vessel is placed inside the bell.
Specifications
Diameter (English)0.43 in.
MaterialBorosilicate Glass
No. per Case1/Cs.
No. per Pack1/Pk.
Height (Metric)85 mm
Height (English)3.34 in.
Diameter (Metric)11 mm
Unit SizeCS