Magnetic Stir Bar Retriever
Actual product may vary
Corning
Magnetic Stir Bar Retriever
For Use With: Stirring Models
Have Questions?
Catalog NumberFor Use With (Equipment)
FIS14-972-50Stirring Models
Catalog number FIS14-972-50
Price (SGD)
114.00
Case
For Use With (Equipment):
Stirring Models
Price (SGD)
114.00
Case
The Corning stir bar retriever contains a strong magnet to remove a stir bar from all glass and plastic containers used in the laboratory. The container must be taken off the magnetic stirrer before using the retriever as the high powered magnet in the stirrer cancels out the one in the retriever. The handle is designed for ergonomic and easy retrieval. -The retriever is available one per case.
Specifications
For Use With (Equipment)Stirring Models
TypeAccessory, Stir Bar Retriever
Unit SizeCase