Epoxy-Coated Test Tube Baskets
Actual product may vary
Black Machine Company
Epoxy-Coated Test Tube Baskets
Epoxy coating is resistant to rust, corrosion, abrasion, solvents, acids, and alkalies
Have Questions?
Catalog NumberDimensions (L x W x H)
FIS14-966-2F10 x 6 x 6 in.
Catalog number FIS14-966-2F
Price (SGD)
-
Dimensions (L x W x H):
10 x 6 x 6 in.
Specifications
AutoclavableAutoclavable
MaterialAluminum
ShapeRectangle
Dimensions (L x W x H)10 x 6 x 6 in.
Height (English)6 in.
Height (Metric)15.2 cm
Unit SizeEach